Merck
  • Good visual outcome after prompt treatment of acanthamoeba keratitis associated with overnight orthokeratology lens wear.
Good visual outcome after prompt treatment of acanthamoeba keratitis associated with overnight orthokeratology lens wear.
Eye & contact lens (2007-11-13)
Victoria W Y Wong, Stanley C C Chi, Dennis S C Lam
ABSTRACT
To describe a patient with a good visual outcome after prompt treatment of Acanthamoeba keratitis as a complication of overnight orthokeratology lens wear. Interventional case report. A 9-year-old boy experienced pain, photophobia, and redness in his right eye 3 days after visiting a swimming pool. He had been wearing overnight orthokeratology lenses for 5 months for the correction of moderate myopia in both eyes. On examination, best-corrected visual acuity in the right eye was 20/40. A diagnosis of Acanthamoeba keratitis with the presence of the classic feature of perineural infiltrates was made. The patient responded well to treatment with polyhexamethylene biguanide and propamidine isethionate (Brolene). Culture of corneal scrapings and contact lens solution showed heavy growth of Acanthamoeba. Treatment was tapered gradually during the next 4 months, and the final best-corrected visual acuity was 20/25. Acanthamoeba keratitis may be a vision-threatening complication associated with overnight orthokeratology lens wear. It is essential for eye care professionals to fully explain and warn parents of the potential downsides that may be associated with orthokeratology. Ophthalmologists should have a high level of suspicion of this complication because prompt diagnosis and treatment can result in good visual outcome.
Product Description
Dibrompropamidine diisetionate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Dibrompropamidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.