Merck
  • Preparation and protein immobilization of magnetic dialdehyde starch nanoparticles.
Preparation and protein immobilization of magnetic dialdehyde starch nanoparticles.
The journal of physical chemistry. B (2013-03-27)
Wensheng Lu, Yuhua Shen, Anjian Xie, Weiqiang Zhang
ABSTRACT
Superparamagnetic Fe3O4 nanoparticles were obtained using a hydrolysis product of starch, i.e., α-d-glucose, as the reducing agent and without any additional stabilizer and dispersant by a facile and green method at mild temperature. Magnetic dialdehyde starch nanoparticles (MDASN) were successfully synthesized with dialdehyde starch (DAS) as wrapper and epichlorohydrin as cross-linker by coembedding method. Bovine serum albumin (BSA) as a model drug was immobilized on the suface of MDASN. The particle size distribution of MDASN was 50-150 nm, and the average size was about 100 nm. The content of aldehyde group in DAS was 59.5%, and the package rate of DAS in MDASN was 33.2%. The loading amount and encapsulation efficiency of MDASN loading BSA were 5.0% and 54.4%, respectively. The saturation magnetization of MDASN at 300 K was 29.5 emu/g without coercivity and remanence. The as-prepared MDASN have not only lots of aldehyde functional groups but also stronger magnetic response, which might have potential applications such as drug carriers and targeted drug release.
Product Description
Supelco
(±)-Epichlorohydrin, analytical standard
Sigma-Aldrich
(±)-Epichlorohydrin, purum, ≥99% (GC)
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis