Skip to Content
Merck
All Photos(1)

Key Documents

02578

Supelco

(±)-Epichlorohydrin

analytical standard

Synonym(s):

(±)-2-(Chloromethyl)oxirane, 1-Chloro-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$387.00

R$387.00


Available to ship onApril 02, 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
R$387.00

About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
Beilstein:
79785
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$387.00


Available to ship onApril 02, 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3.2 (vs air)

vapor pressure

13.8 mmHg ( 21.1 °C)

Assay

≥99.9% (GC)

autoignition temp.

779 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

21 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.438 (lit.)
n20/D 1.438

bp

115-117 °C (lit.)

mp

−57 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClCC1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

BRLQWZUYTZBJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Epichlorohydrin (ECH) is a colorless, volatile and highly reactive organic compound which is utilized as a precursor in the production of glycerol, plastics, drugs and paints.[1][2]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

ECH has been used as a reference standard for the identification of ECH in epoxy-coated cans by oxolane derivatization and gas chromatography−mass spectrometry (GC-MS) in selected ion monitoring (SIM) mode[2] and in drinking water by static headspace (HS) and purge and trap (P&T) coupled to GC equipped with an electron capture detector (GC-ECD).[3]
It may be used as a reference standard for the analysis of (±)-ECH in drinking water by ion chromatography[4] and in effluent water from wastewater treatment plants by HS-solid phase microextraction (HS-SPME) combined with GC-flame ionization detector (GC-FID) and GC-ECD.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

New method for determination of epichlorohydrin in epoxy-coated cans by oxolane derivatization and gas chromatography-mass spectrometry.
Sung, et al.
Journal of Chromatography A, 1201(1), 100-105 (2008)
A convenient method for epichlorohydrin determination in water using headspace-solid-phase microextraction and gas chromatography.
Lasa, et al.
Journal of Chromatographic Science, 44(7), 438-443 (2006)
Determination of epichlorohydrin by ion chromatography.
Sarzanini, et al.
Journal of Chromatography A, 884(1-2), 251-259 (2000)
Static headspace and purge-and-trap gas chromatography for epichlorohydrin determination in drinking water.
Lucentini, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 80(1), 89-98 (2005)
L Cody Smith et al.
Comparative biochemistry and physiology. Part D, Genomics & proteomics, 19, 159-173 (2016-05-09)
The diverse biological effects of xenoestrogens may be explained by their ability to differentially recruit co-regulatory proteins to the estrogen receptor (ER). We employed high-throughput receptor affinity binding and co-regulatory protein recruitment screening assays based on fluorescence polarization and time

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service