Merck
  • Impairment of fear memory consolidation and expression by antihistamines.
Impairment of fear memory consolidation and expression by antihistamines.
Brain research (2012-11-28)
Ayako Nonaka, Fumitaka Masuda, Hiroshi Nomura, Norio Matsuki
ABSTRACT
Antihistamines are widely used to treat allergy symptoms. First-generation antihistamines have adverse effects on the central nervous system (CNS), such as hypnotic and amnesic effects, whereas second-generation antihistamines have poor brain penetration, and therefore, have fewer CNS-related adverse effects. Memory consists of several phases, including acquisition, consolidation, expression, and extinction. It remains unclear whether these phases are affected by antihistamines. We investigated the effects of diphenhydramine, a first-generation antihistamine, and levocetirizine and olopatadine, second-generation antihistamines, on memory phases. Mice were subjected to fear conditioning on day 1 and tested on day 2. Antihistamines were administered before conditioning, immediately after conditioning, or before the test session. Diphenhydramine (30mg/kg) decreased freezing time when administered immediately after conditioning or before the test session. These effects were not attributable to a change in locomotor activity. Levocetirizine (0.1, 1, 10mg/kg) and olopatadine (1, 10, 20mg/kg) had no effects on conditioned fear. We also examined the effect of diphenhydramine and levocetirizine on the expression of an activity-dependent gene associated with the test session. Diphenhydramine, but not levocetirizine, increased Arc transcription in the central nucleus of the amygdala. These data indicate that diphenhydramine, but not levocetirizine or olopatadine, impairs the consolidation and expression of conditioned fear.
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard