Merck
  • Influence of surface wettability on competitive protein adsorption and initial attachment of osteoblasts.
Influence of surface wettability on competitive protein adsorption and initial attachment of osteoblasts.
Biomedical materials (Bristol, England) (2009-06-16)
Jianhua Wei, Toshio Igarashi, Naoto Okumori, Takayasu Igarashi, Takashi Maetani, Baolin Liu, Masao Yoshinari
ABSTRACT
This study investigated the influence of surface wettability on competitive protein adsorption and the initial attachment of osteoblasts. A thin-film coating of hexamethyldisiloxane (HMDSO) and subsequent O(2)-plasma treatment was carried out on substrates with a mirror surface in order to create a wide range of wettabilities. The adsorption behavior of fibronectin (Fn) and albumin (Alb) in both individual and competitive mode, and the initial attachment of mouse osteoblastic cells (MC3T3-E1) over a wide range of wettabilities were investigated. The contact angle of HMDSO coatings without O(2)-plasma treatment against double-distilled water was more than 100 degrees, whereas it dramatically decreased after the O(2)-plasma treatment to almost 0 degrees, resulting in super-hydrophilicity. Individually, Fn adsorption showed a biphasic inclination, whereas Alb showed greater adsorption to hydrophobic surfaces. In the competitive mode, in a solution containing both Fn and Alb, Fn showed greater adsorption on hydrophilic surfaces, whereas Alb predominantly adsorbed on hydrophobic surfaces. The initial attachment of osteoblastic cells increased with an increase in surface wettability, in particular, on a super-hydrophilic surface, which correlated well with Fn adsorption in the competitive mode. These results suggest that Fn adsorption may be responsible for increasing cell adhesion on hydrophilic surfaces in a body fluid or culture media under physiological conditions.
Sigma-Aldrich
Hexamethyldisiloxane, puriss., ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisiloxane, NMR grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Pricing and availability is not currently available.