Skip to Content
Merck
All Photos(1)

Key Documents

52630

Sigma-Aldrich

Hexamethyldisiloxane

puriss., ≥98.5% (GC)

Synonym(s):

HMDSO

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$247.00
500 ML
R$702.00
1 L
R$1,007.00

R$247.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
R$247.00
500 ML
R$702.00
1 L
R$1,007.00

About This Item

Linear Formula:
(CH3)3SiOSi(CH3)3
CAS Number:
Molecular Weight:
162.38
Beilstein:
1736258
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

R$247.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

grade

puriss.

Assay

≥98.5% (GC)

form

liquid

refractive index

n20/D 1.377 (lit.)
n20/D 1.377

bp

101 °C (lit.)

mp

−59 °C (lit.)

density

0.764 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)C

InChI

1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3

InChI key

UQEAIHBTYFGYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HMDSO(hexamethyldisiloxane) is an organosilicon compound. It is a non-toxic, environmentally friendly monomer. It can be used as a precursor to synthesize organosilicon thin films. It serves as a silylating agent for carboxylic acids and alcohols. Additionally, it is used in the preparation of aroyl chlorides. HMDSO is commonly used as a water-repellent.

Application

Hexamethyldisiloxane is used:
  • As a modifying agent in the presence of ethanol/n-hexane solution for the preparation of hydrophobic and hydrophilic sodium silicate-based silica xerogels.
  • To form the dielectric layers in the MIM(metal-insulator-metal) structure that is consisting of graphene quantum dots(GQDs) to prepare two-terminal NVM (non-volatile-memory) devices.

Features and Benefits

Hexamethyldisiloxanehas high thermal stability.

Other Notes

For a facile silylation of carboxylic acids[1][2]; As chloride anion acceptor in the synthesis of sulphinic esters[3]

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
J. Drabowicz
Chemistry Letters (Jpn), 1753-1753 (1981)
Pavel Solař et al.
Scientific reports, 7(1), 8514-8514 (2017-08-19)
Nanoparticles composed of multiple silver cores and a plasma polymer shell (multicore@shell) were prepared in a single step with a gas aggregation cluster source operating with Ar/hexamethyldisiloxane mixtures and optionally oxygen. The size distribution of the metal inclusions as well
H. Matsumoto et al.
Chemistry Letters (Jpn), 1475-1475 (1980)
Edyta Słupek et al.
Materials (Basel, Switzerland), 14(2) (2021-01-10)
The paper presents the screening of 20 deep eutectic solvents (DESs) composed of tetrapropylammonium bromide (TPABr) and glycols in various molar ratios, and 6 conventional solvents as absorbents for removal of siloxanes from model biogas stream. The screening was achieved

Questions

  1. Is PN: 52630 hexamethyldisiloxane dry? How much water in ppm?

    1 answer
    1. This product is not tested for water content. There could be small amounts of water present, but not enough to compromise the 98.5% purity specification. To see all parameters that are tested, please view the specification sheet or sample certificate of analysis, which are linked on the product page.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service