Merck
  • Synthesis, antioxidant properties and radioprotective effects of new benzothiazoles and thiadiazoles.
Synthesis, antioxidant properties and radioprotective effects of new benzothiazoles and thiadiazoles.
Bioorganic & medicinal chemistry (2009-06-09)
Damien Cressier, Caroline Prouillac, Pierre Hernandez, Christine Amourette, Michel Diserbo, Claude Lion, Ghassoub Rima
ABSTRACT
In this work, we report the synthesis and characterization of new compounds derived from benzothiazoles and thiadiazoles. We observed that structural modifications on these skeletons affected the antioxidant activity. Thiol and aminothiol compounds derived from thiadiazoles and benzothiazoles showed an interesting antioxidant property. The radioprotective activity has also been evaluated in mice. Some of these compounds could be good radioprotectors.
Sigma-Aldrich
2-Aminobenzothiazole, 97%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
2-Mercaptobenzothiazole, 97%