Merck
  • 2-Styrylchromones: novel strong scavengers of reactive oxygen and nitrogen species.
2-Styrylchromones: novel strong scavengers of reactive oxygen and nitrogen species.
Bioorganic & medicinal chemistry (2007-07-13)
Ana Gomes, Eduarda Fernandes, Artur M S Silva, Clementina M M Santos, Diana C G A Pinto, José A S Cavaleiro, José L F C Lima
ABSTRACT
2-Styrylchromones are a small group of naturally occurring chromones, vinylogues of flavones (2-phenylchromones). Natural and synthetic 2-styrylchromones have been tested in different biological systems, showing activities with potential therapeutic applications. In particular, the potential and hitherto understudied antioxidant behavior of these compounds has been raised as a matter of interest. Thus the present work consisted in the study of the in vitro scavenging activities for reactive oxygen species (ROS) and reactive nitrogen species (RNS) of various 2-styrylchromone derivatives and structurally similar flavonoids. Some of the studied 2-styrylchromones proved to be extremely efficient scavengers of the different ROS and RNS, showing, in some cases, IC(50)s under 1 microM. The hydroxylation pattern of 2-styrylchromones, especially in the B-ring but also in the A ring, modulates the activity of these compounds, the catecholic derivatives being the most effective scavengers. The styryl pattern also contributes to their observed outstanding antioxidant activity. In conclusion, the scavenging activities for ROS/RNS of 2-styrylchromone derivatives, here shown for the first time, provide novel and most promising compounds to be applied as antioxidants.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
D-Penicillamine, 98-101%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Propyl gallate, powder
Supelco
(R)-(+)-α-Lipoic acid, analytical standard