Merck
  • Sensory Discrimination of Blood and Floral Nectar by Aedes aegypti Mosquitoes.
Sensory Discrimination of Blood and Floral Nectar by Aedes aegypti Mosquitoes.
Neuron (2020-10-14)
Veronica Jové, Zhongyan Gong, Felix J H Hol, Zhilei Zhao, Trevor R Sorrells, Thomas S Carroll, Manu Prakash, Carolyn S McBride, Leslie B Vosshall
ABSTRACT
Blood-feeding mosquitoes survive by feeding on nectar for metabolic energy but require a blood meal to develop eggs. Aedes aegypti females must accurately discriminate blood and nectar because each meal promotes mutually exclusive feeding programs with distinct sensory appendages, meal sizes, digestive tract targets, and metabolic fates. We investigated the syringe-like blood-feeding appendage, the stylet, and discovered that sexually dimorphic stylet neurons taste blood. Using pan-neuronal calcium imaging, we found that blood is detected by four functionally distinct stylet neuron classes, each tuned to specific blood components associated with diverse taste qualities. Stylet neurons are insensitive to nectar-specific sugars and respond to glucose only in the presence of additional blood components. The distinction between blood and nectar is therefore encoded in specialized neurons at the very first level of sensory detection in mosquitoes. This innate ability to recognize blood is the basis of vector-borne disease transmission to millions of people worldwide.
Product Description
Solid-glass beads, borosilicate, diam. 3 mm
Pricing and availability is not currently available.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Millipore
D-(+)-Cellobiose, ≥99.0%, suitable for microbiology, Reducing sugar disaccharide
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.
Millipore
Universal Microbial Limit Test Membrane, white colored, sterile, diameter 70 mm, suitable for microbiology
Pricing and availability is not currently available.