Merck
  • Inhibition of Tumor VEGFR2 Induces Serine 897 EphA2-Dependent Tumor Cell Invasion and Metastasis in NSCLC.
Inhibition of Tumor VEGFR2 Induces Serine 897 EphA2-Dependent Tumor Cell Invasion and Metastasis in NSCLC.
Cell reports (2020-04-30)
Caroline Volz, Sara Breid, Carolin Selenz, Alina Zaplatina, Kristina Golfmann, Lydia Meder, Felix Dietlein, Sven Borchmann, Sampurna Chatterjee, Maike Siobal, Jakob Schöttle, Alexandra Florin, Mirjam Koker, Marieke Nill, Luka Ozretić, Niklas Uhlenbrock, Steven Smith, Reinhard Büttner, Hui Miao, Bingcheng Wang, H Christian Reinhardt, Daniel Rauh, Michael Hallek, Amparo Acker-Palmer, Lukas C Heukamp, Roland T Ullrich
ABSTRACT
Anti-angiogenic treatment targeting vascular endothelial growth factor (VEGF)-VEGFR2 signaling has shown limited efficacy in lung cancer patients. Here, we demonstrate that inhibition of VEGFR2 in tumor cells, expressed in ∼20% of non-squamous non-small cell lung cancer (NSCLC) patients, leads to a pro-invasive phenotype. Drug-induced inhibition of tumor VEGFR2 interferes with the formation of the EphA2/VEGFR2 heterocomplex, thereby allowing RSK to interact with Serine 897 of EphA2. Inhibition of RSK decreases phosphorylation of Serine 897 EphA2. Selective genetic modeling of Serine 897 of EphA2 or inhibition of EphA2 abrogates the formation of metastases in vivo upon VEGFR2 inhibition. In summary, these findings demonstrate that VEGFR2-targeted therapy conditions VEGFR2-positive NSCLC to Serine 897 EphA2-dependent aggressive tumor growth and metastasis. These data shed light on the molecular mechanisms explaining the limited efficacy of VEGFR2-targeted anti-angiogenic treatment in lung cancer patients.
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Dasatinib
Pricing and availability is not currently available.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Pricing and availability is not currently available.