Merck
  • Fumarate Upregulates Surface Expression of ULBP2/ULBP5 by Scavenging Glutathione Antioxidant Capacity.
Fumarate Upregulates Surface Expression of ULBP2/ULBP5 by Scavenging Glutathione Antioxidant Capacity.
Journal of immunology (Baltimore, Md. : 1950) (2020-03-08)
Rikke Illum Høgh, Alec Droujinine, Sofie Hedlund Møller, Stine Dam Jepsen, Maiken Mellergaard, Lars Andresen, Søren Skov
ABSTRACT
Fumarate is a tricarboxylic acid cycle metabolite whose intracellular accumulation is linked to inflammatory signaling and development of cancer. In this study, we demonstrate that endogenous fumarate accumulation upregulates surface expression of the immune stimulatory NK group 2, member D (NKG2D) ligands ULBP2 and ULBP5. In agreement with this, accumulation of fumarate by the therapeutic drug dimethyl fumarate (DMF) also promotes ULBP2/5 surface expression. Mechanistically, we found that the increased ULBP2/5 expression was dependent on oxidative stress and the antioxidants N-acetylcysteine and glutathione (GSH) abrogated ULBP2/5 upregulated by DMF. Fumarate can complex with GSH and thereby exhaust cells of functional GSH capacity. In line with this, inhibition of GSH reductase (GR), the enzyme responsible for GSH recycling, promoted ULBP2/5 surface expression. Loss of the tricarboxylic acid cycle enzyme fumarate hydratase (FH) associates with a malignant form of renal cancer characterized by fumarate accumulation and increased production of reactive oxygen species, highlighting fumarate as an oncometabolite. Interestingly, FH-deficient renal cancer cells had low surface expression of ULBP2/5 and were unresponsive to DMF treatment, suggesting that the fumarate-stimulating ULBP2/5 pathway is abrogated in these cells as an immune-evasive strategy. Together, our data show that ULBP2/5 expression can be upregulated by accumulation of fumarate, likely by depleting cells of GSH antioxidant capacity. Given that DMF is an approved human therapeutic drug, our findings support a broader use of DMF in treatment of cancers and inflammatory conditions.
Sigma-Aldrich
6-Aminonicotinamide, 99%
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Carmustine, ≥98%
Sigma-Aldrich
Sodium propionate, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077 g/mL
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
L-Glutathione oxidized, lyophilized powder
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Dimethyl fumarate, 97%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma