Merck
  • Topology-dependent, bifurcated mitochondrial quality control under starvation.
Topology-dependent, bifurcated mitochondrial quality control under starvation.
Autophagy (2019-06-27)
Yanshuang Zhou, Qi Long, Hao Wu, Wei Li, Juntao Qi, Yi Wu, Ge Xiang, Haite Tang, Liang Yang, Keshi Chen, Linpeng Li, Feixiang Bao, Heying Li, Yaofeng Wang, Min Li, Xingguo Liu
ABSTRACT
Selective elimination of mitochondria by autophagy is a critical strategy for a variety of physiological processes, including development, cell-fate determination and stress response. Although several mechanisms have been identified as responsible for selective degradation of mitochondria, such as the PINK1-PRKN/PARKIN- and receptor-dependent pathways, aspects of the mechanisms and particularly the principles underlying the selection process of mitochondria remain obscure. Here, we addressed a new selection strategy in which the selective elimination of mitochondria is dependent on organellar topology. We found that populations of mitochondria undergo different topological transformations under serum starvation, either swelling or forming donut shapes. Swollen mitochondria are associated with mitochondrial membrane potential dissipation and PRKN recruitment, which promote their selective elimination, while the donut topology maintains mitochondrial membrane potential and helps mitochondria resist autophagy. Mechanistic studies show that donuts resist autophagy even after depolarization through preventing recruitment of autophagosome receptors CALCOCO2/NDP52 and OPTN even after PRKN recruitment. Our results demonstrate topology-dependent, bifurcated mitochondrial recycling under starvation, that is swollen mitochondria undergo removal by autophagy, while donut mitochondria undergo fission and fusion cycles for reintegration. This study reveals a novel morphological selection for control of mitochondrial quality and quantity under starvation.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
5-Hydroxydecanoic acid sodium salt, ≥97% (HPLC)
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Diazoxide
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Target shRNA Control Plasmid DNA, Targets no known genes from any species