Skip to Content
Merck
All Photos(1)

Documents

V900667

Sigma-Aldrich

2,3-Butanediol

Vetec, reagent grade, 98%

Synonym(s):

2,3-Butylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(OH)CH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
969165
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

refractive index

n20/D 1.433 (lit.)

bp

183-184 °C (lit.)

mp

25 °C (lit.)

density

1.002 g/mL at 20 °C (lit.)

SMILES string

CC(O)C(C)O

InChI

1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3

InChI key

OWBTYPJTUOEWEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Plaisant et al.
Free radical biology & medicine, 34(7), 862-872 (2003-03-26)
The pathophysiology of brain lesions associated with cerebral palsy is multifactorial and likely involves excess release of glutamate and excess production of free radicals, among other factors. Theoretically, antioxidants could limit the severity of these brain lesions. Peroxiredoxins are a
Patricia Hermand et al.
The Journal of biological chemistry, 278(7), 4892-4898 (2002-12-13)
ICAM-4 (LW blood group glycoprotein) is an erythroid-specific membrane component that belongs to the family of intercellular adhesion molecules and interacts in vitro with different members of the integrin family, suggesting a potential role in adhesion or cell interaction events
Natsuo Ueda et al.
Neuropharmacology, 48(8), 1079-1085 (2005-05-25)
It is widely accepted that fatty acid amide hydrolase (FAAH) plays a central role in the hydrolysis of anandamide. However, we found a second N-acylethanolamine hydrolase in animal tissues which hydrolyzed anandamide at acidic pH. This "acid amidase" was first
Xiao-Jun Ji et al.
Biotechnology advances, 29(3), 351-364 (2011-01-29)
2,3-butanediol is a promising bulk chemical due to its extensive industry applications. The state-of-the-art nature of microbial 2,3-butanediol production is reviewed in this paper. Various strategies for efficient and economical microbial 2,3-butanediol production, including strain improvement, substrate alternation, and process
Zhi-Long Xiu et al.
Applied microbiology and biotechnology, 78(6), 917-926 (2008-03-06)
1,3-Propanediol and 2,3-butanediol are two promising chemicals which have a wide range of applications and can be biologically produced. The separation of these diols from fermentation broth makes more than 50% of the total costs in their microbial production. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service