Skip to Content
Merck
All Photos(3)

Key Documents

V900594

Sigma-Aldrich

Maleic hydrazide

Vetec, reagent grade, 98%

Synonym(s):

1,2-Dihydro-3,6-pyridazinedione, 3,6-Dihydroxypyridazine, 3,6-Pyridazinediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
Beilstein:
112223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

autoignition temp.

707 °F

mp

299-301 °C (dec.) (lit.)

SMILES string

O=C1NNC(=O)C=C1

InChI

1S/C4H4N2O2/c7-3-1-2-4(8)6-5-3/h1-2H,(H,5,7)(H,6,8)

InChI key

BGRDGMRNKXEXQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Elvia Ruiz Flores et al.
Life sciences, 72(12), 1345-1351 (2003-01-16)
The aim of this investigation was to determine if extracts of Spirulina maxima reduce the genotoxic damage induced by maleic hydrazide (MH) using the Tradescantia biosssay. Two types of extracts from the alga were prepared: an aqueous extract with two
Yanjun Fang et al.
Biosensors & bioelectronics, 24(8), 2323-2327 (2009-03-06)
A novel flow injection chemiluminescence (CL) sensor for the determination of maleic hydrazide (MH) using molecularly imprinted polymer (MIP) as recognition element is reported. The MH-MIP was synthesized by thermal initiated polymerization in methanol, using methacrylic acid (MAA) as functional
Z Swietlińska et al.
Mutation research, 55(1), 15-30 (1978-01-01)
Since 1950, maleic hydrazide (MH) has been introduced into agriculture as a major commercial herbicide and a depressant of plant growth in numerous circumstances such as suppression of sprouting of vegetables and stored food crops, control of sucker growth on
Hassan A Mohamed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(4), 913-917 (2006-02-07)
Interaction of maleic hydrazide (LH(2)) with [Cr(CO)(6)] in air at atmospheric pressure resulted in the formation of the complex [(LH)Cr(mu-O)(2)Cr(LH)] (1). Reaction of LH(2) with [Mo(CO)(6)] in air also gave the complex [(LH(2))O(2)Mo(mu-O)(2)MoO(2)(LH(2))] (2). Under the same conditions, the reaction
Anne-Sophie Foltête et al.
Chemosphere, 85(10), 1624-1629 (2011-09-13)
The Viciafaba root tip micronucleus test is one of the most employed plant genotoxicity assays, and has been used on various types of contaminated materials. This test has been standardized by AFNOR, the French member organization of ISO. However, this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service