Skip to Content
Merck
All Photos(3)

Key Documents

V900431

Sigma-Aldrich

L-Leucine

≥98%, Vetec

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

L-Leucine, Vetec, reagent grade, ≥98%

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

color

white to off-white

mp

>300 °C (lit.)

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William I A Haynes et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(11), 4804-4814 (2013-03-15)
The identification of a hyperdirect cortico-subthalamic nucleus connection highlighted the important role of the subthalamic nucleus (STN) in regulating behavior. However, this pathway was shown primarily from motor areas. Hyperdirect pathways associated with cognitive and motivational cortical regions are particularly
Qian Wang et al.
Journal of the National Cancer Institute, 105(19), 1463-1473 (2013-09-21)
L-type amino acid transporters (LATs) uptake neutral amino acids including L-leucine into cells, stimulating mammalian target of rapamycin complex 1 signaling and protein synthesis. LAT1 and LAT3 are overexpressed at different stages of prostate cancer, and they are responsible for
M Sakamoto et al.
The Journal of clinical endocrinology and metabolism, 56(1), 202-204 (1983-01-01)
Recent studies on the nucleotide sequence of cloned DNA complementary to mRNA for preproenkephalin from bovine adrenal medulla and human pheochromocytoma have revealed that this precursor molecule contains four copies of methionine-enkephalin (Met-Enk) and one copy each of leucine-enkephalin (Leu-Enk)
T Yoshimasa et al.
The Journal of clinical investigation, 69(3), 643-650 (1982-03-01)
To elucidate the physiological and pathophysiological significance of methionine- and leucine-enkephalin (Met-and Leu-enkephalin, respectively) in human sympathoadrenal system, the contents of these peptides in normal human sympathetic nervous system, adrenal medulla, and pheochromocytomas were determined by specific radioimmunoassays combined with
Gaetano Santulli et al.
The Journal of clinical investigation, 125(5), 1968-1978 (2015-04-07)
The type 2 ryanodine receptor (RyR2) is a Ca2+ release channel on the endoplasmic reticulum (ER) of several types of cells, including cardiomyocytes and pancreatic β cells. In cardiomyocytes, RyR2-dependent Ca2+ release is critical for excitation-contraction coupling; however, a functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service