Skip to Content
Merck
All Photos(2)

Key Documents

V900363

Sigma-Aldrich

L-Glutathione oxidized

Vetec, reagent grade, ≥98%

Synonym(s):

GSSG, Glutathiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N6O12S2
CAS Number:
Molecular Weight:
612.63
Beilstein:
1718700
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥98%

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10-,11-,12-/m0/s1

InChI key

YPZRWBKMTBYPTK-BJDJZHNGSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glu-Cys-Gly, Glu-Cys-Gly [Disulfide bridge: 2a-2b]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kondo et al.
American journal of hematology, 26(1), 37-45 (1987-09-01)
The oxidized form of glutathione transport was studied in human erythrocytes in pyrimidine 5'-nucleotidase (P5N) deficiency, a disorder in which the amounts of CTP and UTP in the erythrocytes are elevated. The inhibition of ATP-requiring oxidized glutathione (GSSG) transport by
Roman Kireev et al.
European journal of pharmacology, 701(1-3), 185-193 (2012-12-12)
Fatty livers occur in up to 20% of potential liver donors and increase cellular injury during the ischemia/reperfusion phase, so any intervention that could enable a better outcome of grafts for liver transplantation would be very useful. The effect of
Antonella Caccamo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(3), 906-913 (2013-01-18)
The accumulation of TDP-43 (transactive response DNA-binding protein 43) and its 25 kDa C-terminal fragment (TDP-25) is a hallmark of several neurodegenerative disorders, including frontotemporal lobar degeneration (FTLD-TDP) and amyotrophic lateral sclerosis (ALS). The majority of FTLD-TDP cases are due
Bruce Morgan et al.
Nature chemical biology, 9(2), 119-125 (2012-12-18)
Glutathione is central to cellular redox chemistry. The majority of glutathione redox research has been based on the chemical analysis of whole-cell extracts, which unavoidably destroy subcellular compartment-specific information. Compartment-specific real-time measurements based on genetically encoded fluorescent probes now suggest
Amrit K Sakhi et al.
Journal of chromatography. A, 1104(1-2), 179-189 (2005-12-27)
A method for the simultaneous quantification of reduced and oxidized glutathione in human plasma employing a two-dimensional chromatographic system with parallel porous graphitized carbon (PGC) columns coupled with fluorescence (FLD) and coulometric electrochemical detection (ED) has been developed. Post-sampling oxidation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service