V900341
Adenosine 5′-monophosphate sodium salt
Vetec™, reagent grade, 99%
Synonym(s):
5′-Adenylic acid, A-5′-P, AMP
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H14N5O7P · xNa+ · yH2O
CAS Number:
Molecular Weight:
347.22 (anhydrous free acid basis)
UNSPSC Code:
41106305
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
99%
storage temp.
−20°C
SMILES string
NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@]((COP(O)(O)=O)([H])O3.C.
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shudong Liu et al.
PloS one, 10(5), e0124951-e0124951 (2015-05-02)
Our previous study found that thyroid-stimulating hormone promoted sterol regulatory element-binding protein-2 (SREBP-2) expression and suppressed AMP-activated protein kinase (AMPK) activity in the liver, but it was unclear whether there was a direct link between TSH, AMPK and SREBP-2. Here
Misaki Kawanishi et al.
Biomedical chromatography : BMC, 29(9), 1309-1316 (2015-01-28)
Monitoring of pharmacodynamics in addition to pharmacokinetics is one of strategies to individualize mycophenolate mofetil therapy. The purpose of this study was to develop sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) methods for evaluation of the pharmacokinetics and pharmacodynamics of mycophenolic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service