Skip to Content
Merck
All Photos(3)

Key Documents

V900333

Sigma-Aldrich

2-Thiouracil

Vetec, reagent grade, 99%

Synonym(s):

4-Hydroxy-2-mercaptopyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2OS
CAS Number:
Molecular Weight:
128.15
Beilstein:
112227
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

synthetic

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

mp

>300 °C (lit.)

solubility

1 M NaOH: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

O=C1NC(=S)NC=C1

InChI

1S/C4H4N2OS/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ZEMGGZBWXRYJHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamdouh S Masoud et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 365-372 (2012-10-03)
Mononuclear copper complex [CuL(NH(3))(4)]Cl(2)·0.5H(2)O and three new hetero-metallic complexes: [Cu(2)Ni(L)(2)(NH(3))(2)Cl(2)·6H(2)O] 2H(2)O(,) [Cu(3)Co(L)(4)·8H2O]Cl·4(·)5H(2)O, and [Cu(4)Co(2)Ni(L)(3)(OH)(4)(NH(3))Cl(4)·3H(2)O]4H(2)O where L is 2-thiouracil, were prepared and characterized by elemental analyses, molar conductance, room-temperature magnetic susceptibility, spectral (IR, UV-Vis and ESR) studies and thermal analyses techniques
Leslie Gay et al.
Genes & development, 27(1), 98-115 (2013-01-12)
Transcriptional profiling is a powerful approach for understanding development and disease. Current cell type-specific RNA purification methods have limitations, including cell dissociation trauma or inability to identify all RNA species. Here, we describe "mouse thiouracil (TU) tagging," a genetic and
J Vanden Bussche et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(2), 166-172 (2011-01-26)
Thiouracil belongs to the xenobiotic thyreostats, which are growth-promoting agents illegally used in animal production. Recently it has been reported that thiouracil is suspected to have a natural origin. The European Union of Reference Laboratory guidance paper of 2007 acknowledged
Julie A L Kiebooms et al.
Journal of agricultural and food chemistry, 60(32), 7769-7776 (2012-07-28)
In recent years, the frequent detection of the banned substance thiouracil (TU) in livestock urine has been related to its endogenous formation following consumption of glucosinolate-rich Brassicaceae crops. Besides, TU residues have been recovered in these crops upon plant-derived myrosinase
Raj Kumar Bera et al.
Chemical communications (Cambridge, England), 47(41), 11498-11500 (2011-09-22)
A nonenzymatic method for the selective detection and quantification of serum uric acid (UA) using 2-thiouracil (2-TU) tailored Au nanoparticles is developed. The H-bonding interaction of UA with functionalized Au nanoparticles brings instantaneous visible color change and paves the way

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service