Skip to Content
Merck
All Photos(1)

Key Documents

V900315

Supelco

Ethyl 4-hydroxybenzoate

Vetec, reagent grade, 99%

Synonym(s):

Ethylparaben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101972
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

product line

Vetec

Assay

99%

bp

297-298 °C (lit.)

mp

114-117 °C (lit.)

format

neat

SMILES string

CCOC(=O)c1ccc(O)cc1

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product availableon our online platform ChemisTwin® for NMR. You can use thisdigital equivalent on ChemisTwin® for your sample identityconfirmation and compound quantification (with digital external standard). AnNMR spectrum of this substance can be viewed and an online comparison againstyour sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinshi Oishi
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 42(11), 1845-1849 (2004-09-08)
Parabens are alkyl esters of p-hydroxybenzoic acid widely used as preservatives in foodstuffs, cosmetics toiletries and pharmaceuticals. These compounds are known to exert a weak estrogenic activity in estrogen receptor assays in vitro. In addition butyl and propyl parabens show
Sawsan El Hussein et al.
Experimental dermatology, 16(10), 830-836 (2007-09-12)
Concern is continuously raised about the safety of parabens which are present in most of the cosmetic preparations. In this investigation, methyl-, ethyl-, propyl- and butyl paraben (MP, EP, PP, BP), in a commercial cosmetic lotion, were deposited on human
S Nicoli et al.
Journal of pharmaceutical sciences, 97(11), 4830-4839 (2008-03-05)
This work aims at investigating the nicotinamide (NA)-ethyl-paraben (EP) binary system both in solution and in the solid state. In particular, the apparent EP solubility in water was studied in the presence of different NA concentrations (between 0.28 and 1.64
Christopher Jewell et al.
Toxicology and applied pharmacology, 225(2), 221-228 (2007-09-25)
Parabens are esters of 4-hydroxybenzoic acid and used as anti-microbial agents in a wide variety of toiletries, cosmetics and pharmaceuticals. It is of interest to understand the dermal absorption and hydrolysis of parabens, and to evaluate their disposition after dermal
Lei Wang et al.
Environment international, 59, 27-32 (2013-06-12)
Human exposure to p-hydroxybenzoic acid esters (parabens) is a concern, owing to adverse health effects of these compounds. Parabens are metabolized and eliminated from the human bodies within a few hours of exposure. In this study, for the first time

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service