Skip to Content
Merck
All Photos(3)

Documents

V900311

Sigma-Aldrich

Guanosine

Vetec, reagent grade, ≥98%

Synonym(s):

9-(β-D-Ribofuranosyl)guanine, Guanine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
Beilstein:
625911
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

mp

250 °C (dec.) (lit.)

solubility

formic acid:water (1:1): 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

[H]O[H].NC1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6-,9-/m1/s1

InChI key

NYHBQMYGNKIUIF-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffery T Davis et al.
Chemical Society reviews, 36(2), 296-313 (2007-02-01)
Nature's use of a simple genetic code to enable life's complex functions is an inspiration for supramolecular chemistry. DNA nucleobases carry the key information utilizing a variety of cooperative and non-covalent interactions such as hydrophobic, van der Waals, pi-pi stacking
Cynthia J Burrows et al.
Environmental health perspectives, 110 Suppl 5, 713-717 (2002-11-12)
In vitro work in this laboratory has identified new DNA lesions resulting from further oxidation of a common biomarker of oxidative damage, 8-oxo-7,8-dihydroguanine (OG). The major product of oxidation of OG in a nucleoside, nucleotide, or single-stranded oligodeoxynucleotide using metal
Zoran D Matović et al.
Journal of inorganic biochemistry, 121, 134-144 (2013-02-05)
Novel square-planar palladium(II) complexes with O-N-N-O-type ligands H4mda (H4mda=malamido-N,N'-diacetic acid) and H4obp (H4obp=oxamido-N,N'-di-3-propionic acid) were prepared and characterized. The ligands coordinate to the palladium(II) ion via two pairs of deprotonated ligating atoms with square chelation. A four coordinate, square-planar geometry
Jacob K Grohman et al.
Science (New York, N.Y.), 340(6129), 190-195 (2013-03-09)
RNA chaperones are ubiquitous, heterogeneous proteins essential for RNA structural biogenesis and function. We investigated the mechanism of chaperone-mediated RNA folding by following the time-resolved dimerization of the packaging domain of a retroviral RNA at nucleotide resolution. In the absence
Zhe Li et al.
Nucleic acids research, 42(6), 4068-4079 (2013-12-29)
G-quadruplex-forming oligonucleotides containing modified nucleotide chemistries have demonstrated promising pharmaceutical potential. In this work, we systematically investigate the effects of sugar-modified guanosines on the structure and stability of a (4+0) parallel and a (3+1) hybrid G-quadruplex using over 60 modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service