Skip to Content
Merck
All Photos(3)

Key Documents

V900301

Sigma-Aldrich

N,N′-Methylenebis(acrylamide)

Vetec, reagent grade

Synonym(s):

Bis-acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:

grade

reagent grade

vapor density

5.31 (vs air)

product line

Vetec

form

powder

technique(s)

electrophoresis: suitable

mp

>300 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Lu et al.
ACS nano, 4(12), 7078-7086 (2010-11-19)
Here, we demonstrate that bimetallic Au-Pt nanorods (NRs) can be grown in situ into thermosensitive core-shell microgel particles by a novel two-step approach. In the first step, Au NRs with an average width of 6.6 ± 0.3 nm and length
A J Aho et al.
British journal of anaesthesia, 115(2), 258-266 (2015-07-04)
Concomitantly recorded Bispectral Index® (BIS) and Entropy™ values sometimes show discordant trends during general anaesthesia. Previously, no attempt had been made to discover which EEG characteristics cause discrepancies between BIS and Entropy. We compared BIS and Entropy values, and analysed
Bjørn Bjorvatn et al.
Sleep & breathing = Schlaf & Atmung, 19(4), 1387-1393 (2015-04-10)
The objective of the present study was to investigate the prevalence of insomnia and excessive sleepiness in relation to the presence and severity of obstructive sleep apnea (OSA). The sample consisted of patients referred to a university hospital on suspicion
Barbara R Braams et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(18), 7226-7238 (2015-05-08)
Prior studies have highlighted adolescence as a period of increased risk-taking, which is postulated to result from an overactive reward system in the brain. Longitudinal studies are pivotal for testing these brain-behavior relations because individual slopes are more sensitive for
Jia Liu et al.
Carbohydrate polymers, 94(1), 539-546 (2013-04-03)
To better use wheat straw and minimize its negative impact on environment, a novel semi-interpenetrating polymer networks (semi-IPNs) superabsorbent resin (SAR) composed of wheat straw cellulose-g-poly (potassium acrylate) (WSC-g-PKA) network and linear polyvinyl alcohol (PVA) was prepared by polymerization in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service