Skip to Content
Merck
All Photos(2)

Key Documents

V900286

Sigma-Aldrich

2-Aminopyrimidine

Vetec, reagent grade, 97%

Synonym(s):

2-Pyrimidinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
Beilstein:
107014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

97%

mp

122-126 °C (lit.)

SMILES string

Nc1ncccn1

InChI

1S/C4H5N3/c5-4-6-2-1-3-7-4/h1-3H,(H2,5,6,7)

InChI key

LJXQPZWIHJMPQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Ermolat'ev et al.
Molecular diversity, 15(2), 491-496 (2010-08-27)
An efficient microwave-assisted one-pot two-step protocol was developed for the construction of disubstituted 2-amino-1H-imidazoles. This process involves the sequential formation of 2,3-dihydro-2-hydroxyimidazo[1,2-a]pyrimidinium salts from readily available 2-aminopyrimidines and α-bromoketones, followed by cleavage of the pyrimidine ring with hydrazine.
S A Abdel-Latif et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(3-4), 950-957 (2006-11-07)
The formation constants of some transition metal ions Cr(III), Mn(II), Fe(III), Ni(II) and Cu(II) binary complexes containing Schiff bases resulting from condensation of salicylaldehyde with aniline (I), 2-aminopyridine (II), 4-aminopyridine (III) and 2-aminopyrimidine (IV) were determined pH-metrically in ethanolic medium
Rogier A Smits et al.
Drug discovery today, 14(15-16), 745-753 (2009-05-30)
The search for new and potent histamine H4 receptor ligands is leading to a steadily increasing number of scientific publications and patent applications. Several interesting and structurally diverse compounds have been found, but fierce IP competition for a preferred 2-aminopyrimidine
Teodor Silviu Balaban et al.
Journal of the American Chemical Society, 125(14), 4233-4239 (2003-04-03)
The 2-aminopyrimidin-5-yl ligand is revealed to be a promising candidate for the construction of supramolecular porphyrin arrays with broad absorption bands for efficient light-harvesting. 10-Mono- and 10,20-di(2-aminopyrimidin-5-yl) derivatives of 5,15-bis(3,5-di-tert-butylphenyl)porphyrin have been synthesized in high yield. Their Zn(II) salts show
Erick A Lindsey et al.
Organic & biomolecular chemistry, 10(13), 2552-2561 (2012-02-04)
An efficient synthetic route to a series of substituted 2-aminopyrimidine (2-AP) derivatives has been developed. Subsequent biofilm screening has allowed comparison between the biological activity of these new derivatives and that of the 2-aminoimidazole class of anti-biofilm compounds. Several derivatives

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service