Skip to Content
Merck
All Photos(2)

Documents

V900209

Sigma-Aldrich

Sodium tetraborate decahydrate

Vetec, reagent grade, 99%

Synonym(s):

Borax decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

pH

9.2 (10 g/L)

density

1.73 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

CDMADVZSLOHIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Smartphone-based digital image colorimetry for the determination of total capsaicinoid content in chili pepper extracts.: Utilizing 2,6-Dichloroquinone-4-chloroimide in an innovative smartphone-based colorimetric analysis, this study presents a cost-effective and accessible method for quantifying capsaicinoids in chili extracts, enhancing food quality control and research in capsaicinoid-related health benefits (Vakh et al., 2024).

  • Multispectral and Molecular Docking Studies Reveal Potential Effectiveness of Antidepressant Fluoxetine by Forming Π-Acceptor Complexes.: This research explores the interaction of fluoxetine with 2,6-Dichloroquinone-4-chloroimide, demonstrating the compound′s role in enhancing the drug′s antidepressant effects through molecular docking studies, suggesting new therapeutic strategies (Gaber et al., 2022).


Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunice F S Vieira et al.
Biomacromolecules, 9(4), 1195-1199 (2008-03-19)
Oxidized alginate (ADA) and oxidized alginate blended with chitosan (ADA-Chit) were prepared in the presence of borax and CaCl 2, and their interactions with an antifolate drug, pyrimethamine (PYR), have been investigated. Tablets with a mean diameter of 1.2 +/-
Hasan Turkez
Journal of applied toxicology : JAT, 28(5), 658-664 (2007-11-21)
Titanium dioxide (TiO(2)) is a potential carcinogenic/mutagenic agent although it is used in many areas including medical industries and cosmetics. Boron (as boric acid and borax) has also well-described biological effects and therapeutic benefits. In a previous study, sister-chromatid exchanges
Jian-Feng He et al.
Journal of chromatography. A, 1218(24), 3816-3821 (2011-05-14)
The paper presents an on-line transient moving chemical reaction boundary (MCRB) method for simply but efficiently stacking analytes in capillary electrophoresis (CE). The CE technique was developed for a rapid determination of fumaric and maleic acid. Based on the theory
Xuan Wei et al.
Journal of chromatography. A, 1281, 148-154 (2013-02-13)
With an increasing concern on food safety, fast screening of residues of widespread herbicides becomes necessary. Herein we report a microchip electrophoresis system with laser induced fluorescence (LIF) detection for rapid and sensitive analysis of glyphosate (GLYP) and glufosinate (GLUF)
Qingzhou Cui et al.
The Analyst, 134(5), 875-880 (2009-04-22)
We developed a photonic crystal sensing method for diol containing species such as carbohydrates based on a poly(vinyl alcohol) (PVA) hydrogel containing an embedded crystalline colloidal array (CCA). The polymerized CCA (PCCA) diffracts visible light. We show that in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service