V900105
Ammonium oxalate monohydrate
Vetec™, reagent grade, 98%
Synonym(s):
Oxalic acid diammonium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
142.11
Beilstein:
3759830
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
98%
grade:
reagent grade
form:
crystals, granules or powder
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
98%
form
crystals, granules or powder
pH
6.0-7.0 (25 °C, 14.2 g/L)
mp
133 °C (dec.) (lit.)
SMILES string
OC(C(O)=O)=O.N.O.N
InChI
1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2
InChI key
MSMNVXKYCPHLLN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yuancun Qiao et al.
The journal of physical chemistry letters, 6(14), 2755-2760 (2015-08-13)
Negative linear compressibility (NLC) is a relatively uncommon phenomenon and rarely studied in organic systems. Here we provide the direct evidence of the persistent NLC in organic mineral ammonium oxalate monohydrate under high pressure using synchrotron X-ray powder diffraction, Raman
Electrochemiluminescence as a tool for microscopy at the nanoscale.
Lee A, et al.
Proceedings of SPIE, 13, 13-20 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service