Skip to Content
Merck
All Photos(2)

Documents

V900022

Sigma-Aldrich

Rubidium chloride

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

fine crystals (may contain lumps)

mp

715 °C (lit.)

density

2.8 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[Rb+]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

InChI key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luuk Mestrom et al.
Applied and environmental microbiology, 85(8) (2019-02-10)
LeLoir glycosyltransferases are important biocatalysts for the production of glycosidic bonds in natural products, chiral building blocks, and pharmaceuticals. Trehalose transferase (TreT) is of particular interest since it catalyzes the stereo- and enantioselective α,α-(1→1) coupling of a nucleotide sugar donor
Jie-Ling Xie et al.
Nanoscale, 11(14), 6719-6726 (2019-03-23)
The development of nontoxic and stable all-inorganic perovskite nanocrystals (NCs) represents a major challenge to unlock widely practical applications in photoelectric fields. It is an effective strategy to discover nontoxic metal perovskite nanocrystals (NCs) with excellent optical properties. In addition
F Stevens et al.
Physical chemistry chemical physics : PCCP, 7(2), 240-249 (2005-01-21)
An extensive level of theory study is performed on diatomic chalcogen defects in alkali halide lattices by density functional theory methods. A variety of exchange correlation functionals and basis sets are used for the calculation of electron paramagnetic resonance (EPR)
Christophe Danelon et al.
Biophysical chemistry, 104(3), 591-603 (2003-08-14)
Ion current through single outer membrane protein F (OmpF) trimers was recorded and compared to molecular dynamics simulation. Unidirectional insertion was revealed from the asymmetry in channel conductance. Single trimer conductance showed particularly high values at low symmetrical salt solution.
Yucheng Jiang et al.
Carbohydrate research, 341(2), 262-269 (2005-12-07)
The Gibbs energy interaction parameters of RbCl with some monosaccharides (D-glucose, D-galactose, D-xylose, and D-arabinose) in water, g(ES), were obtained from electromotive force (emf) measurements of the electrochemical cell without liquid junction and containing two ion-selective electrodes (ISE): K-ISEmid R:RbCl(m(E))mid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service