Skip to Content
Merck
All Photos(1)

Key Documents

V900006

Sigma-Aldrich

Biphenyl

Vetec, reagent grade, 99%

Synonym(s):

1,1-Biphenyl, Diphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21
Beilstein:
1634058
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

5.31 (vs air)

vapor pressure

9.46 mmHg ( 115 °C)

product line

Vetec

Assay

99%

autoignition temp.

1004 °F

expl. lim.

0.6 %, 111 °F
5.8 %, 166 °F

bp

255 °C (lit.)

mp

68-70 °C (lit.)

SMILES string

c1ccc(cc1)-c2ccccc2

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianming Zhang et al.
Environmental science. Processes & impacts, 17(12), 2006-2012 (2015-11-26)
Passive air samplers (PASs) are simple, versatile devices that are increasingly used to determine the concentrations of semivolatile organic compounds (SVOCs) in the atmosphere. Using PAS and interpreting PAS-derived data with confidence requires a detailed understanding of the factors that
Wei Wang et al.
Environment international, 83, 183-191 (2015-07-16)
Tetrabromobisphenol A (TBBPA) and eight bisphenol analogues (BPs) including bisphenol A (BPA) were determined in 388 indoor (including homes and microenvironments) dust samples collected from 12 countries (China, Colombia, Greece, India, Japan, Kuwait, Pakistan, Romania, Saudi Arabia, South Korea, U.S.
Shi-Rong Liu et al.
Organic letters, 15(4), 878-881 (2013-02-05)
A BODIPY-based fluorescent probe, HCSe, has been successfully developed for the rapid detection of hypochlorous acid based on the specific HOCl-promoted oxidation of diphenyl selenide in response to the amount of HOCl. Confocal fluorescence microscopy imaging using RAW264.7 cells showed
Sheng-Hao Hsu et al.
Acta biomaterialia, 8(11), 4151-4161 (2012-07-31)
Low-shrinkage resin-based photocurable liquid crystalline epoxy nanocomposite has been investigated with regard to its application as a dental restoration material. The nanocomposite consists of an organic matrix and an inorganic reinforcing filler. The organic matrix is made of liquid crystalline
Xiaofeng Chen et al.
The Journal of organic chemistry, 77(17), 7572-7578 (2012-08-07)
A highly efficient cross-coupling of diarylborinic acids and anhydrides with aryl chlorides and bromides has been effected by using a palladium catalyst system co-supported by a strong σ-donor N-heterocyclic carbene (NHC), N,N'-bis(2,6-diisopropylphenyl) imidazol-2-ylidene, and a strong π-acceptor phosphite, triphenylphosphite, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service