Skip to Content
Merck
All Photos(4)

Key Documents

V800252

Sigma-Aldrich

Mercury(II) chloride

AR, ≥99%

Synonym(s):

Mercuric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgCl2
CAS Number:
Molecular Weight:
271.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

vapor pressure

1.3 mmHg ( 236 °C)

product line

Vetec

Assay

≥99%

form

crystalline powder

mp

277 °C (lit.)

SMILES string

Cl[Hg]Cl

InChI

1S/2ClH.Hg/h2*1H;/q;;+2/p-2

InChI key

LWJROJCJINYWOX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kshitiz Tyagi et al.
Nucleic acids research, 43(9), 4701-4712 (2015-04-15)
Maintenance of protein quality control has implications in various processes such as neurodegeneration and ageing. To investigate how environmental insults affect this process, we analysed the proteome of yeast continuously exposed to mild heat stress. In agreement with previous transcriptomics
Michael Gülden et al.
Toxicology, 332, 41-51 (2013-12-03)
In vitro generated data on toxic potencies are generally based on nominal concentrations. However, cellular and extracellular binding and elimination processes may reduce the available free fraction of a compound. Then, nominal effective concentrations do not represent appropriate measures of
Andrés Cruz Hernández et al.
Metallomics : integrated biometal science, 5(4), 398-403 (2013-03-27)
RNA processing is an essential pathway in the regulation of genetic expression in the cell. In this work, Bacillus subtilis was used to understand the effects of mercury on the mechanism of tRNA metabolism. The CVAAS (cold vapor atomic absorption
Susan C Fitzer et al.
Scientific reports, 4, 6218-6218 (2014-08-29)
Ocean acidification is altering the oceanic carbonate saturation state and threatening the survival of marine calcifying organisms. Production of their calcium carbonate exoskeletons is dependent not only on the environmental seawater carbonate chemistry but also the ability to produce biominerals
Masaro Akai et al.
Journal of bacteriology, 194(24), 6828-6836 (2012-10-09)
The moderately halotolerant cyanobacterium Synechocystis sp. strain PCC 6803 contains a plasma membrane aquaporin, AqpZ. We previously reported that AqpZ plays a role in glucose metabolism under photomixotrophic growth conditions, suggesting involvement of AqpZ in cytosolic osmolarity homeostasis. To further

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service