Skip to Content
Merck
All Photos(1)

Key Documents

V800247

Sigma-Aldrich

Manganese(II) chloride

AR, ≥99%

Synonym(s):

Manganese dichloride, Sacchite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99%
grade:
AR
form:
crystalline powder

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

5.0-6.0 (5%)

mp

652 °C (lit.)

density

2.98 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.01%
Ca: ≤0.03%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.005%
Na: ≤0.05%
Ni: ≤0.002%
Zn: ≤0.005%
heavy metals (as Pb): ≤0.0005%

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tora Sund Morken et al.
Pediatric research, 73(2), 171-179 (2012-11-24)
Hypoxia-ischemia (HI) induces delayed inflammation and long-term gray and white matter brain injury that may be altered by hyperoxia. HI and 2 h of hyperoxia (100% O2) or room air (21% O2) in 7-d-old (P7) rats were studied by magnetic resonance
M R Sepúlveda et al.
Contrast media & molecular imaging, 7(4), 426-434 (2012-06-01)
Manganese is a vital element and cofactor of many key enzymes, but it is toxic at high levels, causing pronounced disturbances in the mammalian brain. Magnetic resonance imaging (MRI) studies using manganese ions as a paramagnetic contrast agent are often
Oliviero L Gobbo et al.
Contrast media & molecular imaging, 7(6), 542-546 (2012-09-20)
Lung parenchyma remains one of the most difficult tissues to be imaged by means of magnetic resonance imaging (MRI). Several MRI techniques are routinely used for lung imaging. However, manganese-enhancement MRI (MEMRI) technique has not been associated with pulmonary MRI.
Elizabeth Greally et al.
Journal of cardiovascular magnetic resonance : official journal of the Society for Cardiovascular Magnetic Resonance, 15, 4-4 (2013-01-18)
Manganese-enhanced cardiovascular magnetic resonance (MECMR) can non-invasively assess myocardial calcium influx, and calcium levels are known to be elevated in muscular dystrophy cardiomyopathy based on cellular studies. Left ventricular functional studies and MECMR were performed in mdx mice (model of
Asad A Aboud et al.
Neurotoxicology, 33(6), 1443-1449 (2012-10-27)
Manganese (Mn) is an environmental risk factor for Parkinson's disease (PD). Recessive inheritance of PARK2 mutations is strongly associated with early onset PD (EOPD). It is widely assumed that the influence of PD environmental risk factors may be enhanced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service