Skip to Content
Merck
All Photos(1)

Key Documents

V800150

Sigma-Aldrich

Dibutyl phthalate

LR, ≥98%

Synonym(s):

n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
1914064
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
bp:
340 °C (lit.)
vapor pressure:
1 mmHg ( 147 °C)

grade

LR

vapor density

9.6 (vs air)

vapor pressure

1 mmHg ( 147 °C)

product line

Vetec

Assay

≥98%

form

liquid

autoignition temp.

756 °F

expl. lim.

0.47 %, 236 °F

dilution

(for analytical testing)

refractive index

n20/D 1.492 (lit.)

pH

7 (20 °C, 10 mg/L)

bp

340 °C (lit.)

mp

−35 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

367.7 °F - open cup

Flash Point(C)

186.5 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alin C Dirtu et al.
Environment international, 59, 344-353 (2013-07-31)
Human exposure to chemicals commonly encountered in our environment, like phthalates, is routinely assessed through urinary measurement of their metabolites. A particular attention is given to the specific population groups, such as obese, for which the dietary intake of environmental
Harpreet Bhatia et al.
Environmental toxicology and chemistry, 32(10), 2335-2344 (2013-06-14)
Di-n-butyl phthalate (DnBP) is an industrial pollutant with antiandrogenic effects reported in male mammals and fish. Little research has been done on the endocrine effects of DnBP in female fish. The present study investigated the changes in ovarian histology and
Harpreet Bhatia et al.
Aquatic toxicology (Amsterdam, Netherlands), 149, 103-115 (2014-03-01)
Phthalic acid esters (PAEs) are a class of synthetic industrial chemicals commonly found in the aquatic environment worldwide. PAEs have been recognised as anti-androgens in male mammals but little is known about their endocrine disrupting effects in fish. This study
Sarah Moody et al.
Endocrinology, 154(9), 3460-3475 (2013-06-15)
Phthalates are plasticizers with widespread industrial, domestic, and medical applications. Epidemiological data indicating increased incidence of testicular dysgenesis in boys exposed to phthalates in utero are reinforced by studies demonstrating that phthalates impair fetal rodent testis development. Because humans are
Yuichiro Yano et al.
Journal of the American College of Cardiology, 65(4), 327-335 (2015-01-31)
Isolated systolic hypertension (ISH), defined as systolic blood pressure (SBP) ≥140 mm Hg and diastolic blood pressure (DBP) <90 mm Hg, in younger and middle-aged adults is increasing in prevalence. The aim of this study was to assess the risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service