Skip to Content
Merck
All Photos(1)

Key Documents

V800144

Sigma-Aldrich

Diisopropyl ether

LR, ≥98%

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
Assay:
≥98%
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)

grade

LR

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

product line

Vetec

Assay

≥98%

form

liquid

autoignition temp.

827 °F

expl. lim.

1-21 %, 100 °F

refractive index

n20/D 1.367 (lit.)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.725 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fiorella Belpoggi et al.
Annals of the New York Academy of Sciences, 982, 70-86 (2003-02-04)
Tert-amyl-methyl ether (TAME) was administered by gavage in extra virgin olive oil solution at concentrations of 750, 250, or 0 mg/kg bw to groups of 100 male and 100 female Sprague-Dawley rats 8 weeks old at the start of the
A Hickel et al.
Biotechnology and bioengineering, 65(4), 425-436 (1999-10-03)
A novel recycle reactor has been designed to determine the interfacial activity of hydroxynitrile lyase in a diisopropyl ether (DIPE)/water two-phase system. The reactor provides a known interfacial area. Enzyme activity toward mandelonitrile cleavage is continuously measured in the reactor
I Linhart et al.
Toxicology and applied pharmacology, 136(1), 155-160 (1996-01-01)
Biotransformation of acrolein (ACR) was studied in vivo in the rat following inhalation and ip administration. The major and minor urinary metabolites were 3-hydroxypropylmercapturic acid (HPMA) and 2-carboxyethylmercapturic acid (CEMA), respectively. Male Wistar rats were exposed to ACR, 23, 42
P Adlercreutz
Biochimica et biophysica acta, 1163(2), 144-148 (1993-05-13)
Horse liver alcohol dehydrogenase and alpha-chymotrypsin were deposited on a porous support material, Celite. After equilibration at a well-defined water activity, the catalytic activity was measured with diisopropyl ether as reaction medium. The effects of the presence of polyols and
B E Cham et al.
Journal of virological methods, 137(1), 160-163 (2006-07-01)
Many viruses including HIV, hepatitis C and hepatitis B, have an outer lipid envelope which maintains inserted viral peptides in the "correct" functional conformation and orientation. Disruption of the lipid envelope by most solvents destroys infectivity and often results in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service