Journal of photochemistry and photobiology. B, Biology, 90(1), 1-7 (2007-11-21)
The interaction between bromocresol purple (BCP) and bovine serum albumin (BSA)/porcine serum albumin (PSA) was investigated both in the absence and presence of bilirubin (BR) using absorption/absorption difference spectroscopy. A significant red shift in the absorption maxima of BCP accompanied
Determination of thermodynamic parameters of water oxidation at the photosystem II (PSII) manganese complex is a major challenge. Photothermal beam deflection (PBD) spectroscopy determines enthalpy changes (ΔH) and apparent volume changes which are coupled with electron transfer in the S-state
Nutrition (Burbank, Los Angeles County, Calif.), 25(10), 1006-1010 (2009-06-03)
Hypoalbuminemia is known to occur in critically ill patients and is associated with increased mortality. Severe hypoalbuminemia is defined in the literature as serum albumin levels lower than 24 g/L. Albumin levels were measured in our laboratory using the bromocresol
Journal of dairy science, 89(7), 2459-2464 (2006-06-15)
The MicroFoss method was evaluated for its effectiveness as an indicator of fluid milk shelf life. Half-gallon, 2% fat fluid milk samples (n = 90) were obtained from a milk processing plant on 3 occasions postpasteurization and evaluated for shelf
Journal of hazardous materials, 166(2-3), 701-705 (2009-01-13)
A simple, selective, and sensitive kinetic spectrophotometric method is described for the determination of trace amounts of phenylhydrazine, which is based on its inhibition effect on the reaction between meta cresol purple (MCP) and periodate in the presence of bromide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.