V800030
Ammonium hydroxide solution
AR, ~25% NH3
Synonym(s):
Ammonia aqueous, Ammonia water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Recommended Products
grade
AR
product line
Vetec™
form
liquid
expl. lim.
27 %
concentration
~25% NH3
density
0.9 g/mL at 25 °C (lit.)
SMILES string
[NH4+].[OH-]
InChI
1S/H3N.H2O/h1H3;1H2
InChI key
VHUUQVKOLVNVRT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each
Enrique Cequier et al.
Analytica chimica acta, 845, 98-104 (2014-09-10)
Organophosphate triesters are common flame retardants used in a wide variety of consumer products from which they can migrate and pollute the indoor environment. Humans may thus be continuously exposed to several organophosphate triesters which might be a risk for
Dominic J Hare et al.
ACS chemical neuroscience, 6(3), 398-402 (2015-01-15)
Plasma iron levels are decreased in Alzheimer's disease (AD) and associated with an idiopathic anemia. We examined iron-binding plasma proteins from AD patients and healthy controls from the Australian Imaging, Biomarkers and Lifestyle (AIBL) Flagship Study of Ageing using size
Marilia Freitas Calmon et al.
Colloids and surfaces. B, Biointerfaces, 100, 177-184 (2012-07-07)
Magnetic iron oxide nanoparticles (magnetite) (MNPs) were prepared using different organic and inorganic bases. Strong inorganic base (KOH) and organic bases (NH(4)OH and 1,4-diazabicyclo[2.2.2]octane (DABCO)) were used in the syntheses of the MNPs. The MNPs were characterized by X-ray diffraction
Aurélie Periat et al.
Journal of chromatography. A, 1356, 211-220 (2014-07-16)
In this study, the influence of electrospray ionization (ESI) source design on the overall sensitivity achieved in hydrophilic interaction chromatography (HILIC) and reversed phase liquid chromatography (RPLC), was investigated. State-of-the-art triple quadrupole mass analyzers from AB Sciex, Agilent Technologies and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service