Skip to Content
Merck
All Photos(1)

Key Documents

1570439

USP

2-Propanol System Suitability

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Isopropanol Suitability Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product 1570439 is a customer specific product and is not available to be sold on the internet. Contact Technical Service

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

It is intended to be used as a USP reference standard in specified quality tests and assays.
Also used to prepare system suitability solution for assay and volatile impurities analysis using gas chromatography-flame ionization detector (GC-FID) as described in the following United States Pharmacopeia (USP) monographs:
  • Isopropyl alcohol
  • Azeotropic Isopropyl Alcohol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isopropyl alcohol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(3), 2428-2428 (2022)
Azeotropic Isopropyl Alcohol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(5), 2430-2430 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service