1496802
USP
Parachlorophenol
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
4-Chlorophenol
About This Item
Recommended Products
grade
pharmaceutical primary standard
vapor pressure
1 mmHg ( 49.8 °C)
API family
parachlorophenol
manufacturer/tradename
USP
bp
220 °C (lit.)
mp
42-45 °C
density
1.306 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Oc1ccc(Cl)cc1
InChI
1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H
InChI key
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monograph such as:
- Triclosan
- Chloroxylenol
Analysis Note
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
249.8 °F - closed cup
Flash Point(C)
121 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service