Skip to Content
Merck
All Photos(1)

Documents

1487007

USP

Oxymetholone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

17β-Hydroxy-2-(hydroxymethylene)-17-methyl-5α-androstan-3-one, Anasterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxymetholone

manufacturer/tradename

USP

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CC[C@]3([H])C(CC[C@@]4(C)[C@@]3([H])CC[C@]4(C)O)[C@@]1(C)C\C(=C\O)C(=O)C2

InChI

1S/C21H32O3/c1-19-11-13(12-22)18(23)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,22,24H,4-11H2,1-3H3/b13-12-/t14-,15+,16?,17-,19-,20-,21-/m0/s1

InChI key

ICMWWNHDUZJFDW-ZDBQEZQZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Oxymetholone in Muscle Growth Research: A study on the synergistic blend of Garcinia mangostana and Cinnamomum tamala suggests potential for enhancing myogenic differentiation and mitochondrial biogenesis, similar to effects observed with Oxymetholone in promoting muscle growth and strength (Sinha et al., 2023).

  • Oxymetholone in Combination Therapy: Long-term combination therapy with Metformin and Oxymetholone in a Fanconi Anemia mouse model shows significant promise for treating this genetic disorder, suggesting new therapeutic pathways involving this potent anabolic steroid (Dorrell et al., 2023).

  • Cost-effectiveness of Oxymetholone: Real-world analysis in Thailand compares the cost-effectiveness of Oxymetholone versus Rabbit-Antithymocyte Globulin for treating acquired aplastic anemia, providing valuable insights into healthcare resource allocation (Penthinapong et al., 2023).

  • Oxymetholone and Testicular Toxicity: Research investigates the effects of Platelet-Rich Plasma on Oxymetholone-induced testicular toxicity, contributing to understanding its adverse effects and potential mitigation strategies (Saba et al., 2023).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service