1374361
USP
Magnesium sulfate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Magnesium sulfate heptahydrate, Epsom salts
About This Item
Recommended Products
grade
pharmaceutical primary standard
vapor density
<0.01 (vs air)
vapor pressure
<0.1 mmHg ( 20 °C)
API family
magnesium sulfate
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
InChI
1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2
InChI key
WRUGWIBCXHJTDG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service