Skip to Content
Merck
All Photos(1)

Key Documents

1274630

USP

Flunixin Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methyl-3-(trifluoromethyl)aniline, 2-Amino-6-(trifluoromethyl)toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(CF3)NH2
CAS Number:
Molecular Weight:
175.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Product 1274630 is a customer specific product and is not available to be sold on the internet. Contact Technical Service

grade

pharmaceutical primary standard

API family

flunixin

manufacturer/tradename

USP

mp

38-42 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1c(N)cccc1C(F)(F)F

InChI

1S/C8H8F3N/c1-5-6(8(9,10)11)3-2-4-7(5)12/h2-4H,12H2,1H3

InChI key

TWLDBACVSHADLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flunixin Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Flunixin Meglumine

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

205.0 °F - closed cup

Flash Point(C)

96.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service