Skip to Content
Merck
All Photos(1)

Documents

TA9H97BAF1A0

Targetmol Chemicals Inc.

Buserelin Acetate (57982-77-1 free base)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
1299.49

Iupac Name

(S)-1-((3S,6S,9S,12S,15R,18S,21S)-3-((1H-imidazol-5-yl)methyl)-6-((1H-indol-3-yl)methyl)-15-(tert-butoxymethyl)-21-(3-guanidinopropyl)-12-(4-hydroxybenzyl)-9-(hydroxymethyl)-18-isobutyl-1,4,7,10,13,16,19-heptaoxo-1-((S)-5-oxopyrrolidin-2-yl)-2,5,8,11,14,17,20-heptaazadocosane)-N-ethylpyrrolidine-2-carboxamide acetate

Inchi Code

1S/C60H86N16O13.C2H4O2/c1-7-64-57(87)48-15-11-23-76(48)58(88)41(14-10-22-65-59(61)62)69-51(81)42(24-33(2)3)70-56(86)47(31-89-60(4,5)6)75-52(82)43(25-34-16-18-37(78)19-17-34)71-55(85)46(30-77)74-53(83)44(26-35-28-66-39-13-9-8-12-38(35)39)72-54(84)45(27-36-29-63-32-67-36)73-50(80)40-20-21-49(79)68-40;1-2(3)4/h8-9,12-13,16-19,28-29,32-33,40-48,66,77-78H,7,10-11,14-15,20-27,30-31H2,1-6H3,(H,63,67)(H,64,87)(H,68,79)(H,69,81)(H,70,86)(H,71,85)(H,72,84)(H,73,80)(H,74,83)(H,75,82)(H4,61,62,65);1H3,(H,3,4)/t40-,41-,42-,43-,44-,45-,46-,47+,48-;/m0./s1

InChI key

PYMDEDHDQYLBRT-DRIHCAFSSA-N

storage temp.

-80/-20

Purity

99.23%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Buserelin Acetate (57982-77-1 free base)

MSDS

https://www.targetmol.com/compound/Buserelin Acetate (57982-77-1 free base)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service