Skip to Content
Merck
All Photos(1)

Documents

Y0002117

Tetracaine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

tetracaine

manufacturer/tradename

EDQM

storage temp.

2-8°C

SMILES string

CCCCNc1ccc(cc1)C(=O)OCCN(C)C

InChI

1S/C15H24N2O2/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3/h6-9,16H,4-5,10-12H2,1-3H3

InChI key

GKCBAIGFKIBETG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Topical ophthalmic anesthetic; used for spinal anesthesia

Biochem/physiol Actions

Blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service