Y0002116
Propyl gallate for ID and assay
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Propyl gallate, 3,4,5-Trihydroxybenzoic acid propyl ester
About This Item
Recommended Products
API family
propyl gallate
manufacturer/tradename
EDQM
mp
146-149 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCOC(=O)c1cc(O)c(O)c(O)c1
InChI
1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3
InChI key
ZTHYODDOHIVTJV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Packaging
Reconstitution
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
368.6 °F - closed cup
Flash Point(C)
187 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service