SU860178
Short thread screw cap, 9 mm thread, with liner
Synonym(s):
Short thread screw cap, 9 mm thread, with liner
About This Item
Recommended Products
material
beige silicone
blue polypropylene screw cap (open-top)
white PTFE/silicone septum (with slit)
feature
Ultrabond
hardness
45 (Durometer: shore A)
packaging
pkg of 1 × 100 ea (SU860178)
availability
not available in USA
thickness
1.3 mm
fitting
for 9 mm short thread
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service