Skip to Content
Merck
All Photos(1)

Key Documents

S30000S

High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 4965 mPa.s (60 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
R$996.00

R$996.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
R$996.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$996.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

4965 mPa.s(60 °C)

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon High Temperature Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service