Skip to Content
Merck
All Photos(1)

Key Documents

PS676

Methamidophos

analytical standard

Synonym(s):

O,S-Dimethyl phosphoramidothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H8NO2PS
CAS Number:
Molecular Weight:
141.13
Beilstein:
8137714
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-676

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COP(N)(=O)SC

InChI

1S/C2H8NO2PS/c1-5-6(3,4)7-2/h1-2H3,(H2,3,4)

InChI key

NNKVPIKMPCQWCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methamidophos is a nonselective type of organophosphorus insecticide and a potent acetylcholinesterase inhibitor, which is commonly used in controlling a broad spectrum of pest insects such as chewing and sucking insects and spider mites on citrus fruits, ornamental plants, stone fruits and other intensive agricultural crops.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

413.6 °F - closed cup

Flash Point(C)

212 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titanium dioxide mediated photocatalytic degradation of methamidophos in aqueous phase.
Wei L, et al.
Journal of Hazardous Materials, 164(1), 154-160 (2009)
Carla S Lima et al.
Neurotoxicology, 32(6), 718-724 (2011-08-30)
Epidemiologic studies describe a potential risk of depression and suicide in farm workers exposed to organophosphates (OPs). In a previous study we observed an increase in depressive-like behavior in adult mice exposed to the OP pesticide methamidophos. Considering the association
Guilherme L Emerick et al.
Toxicology, 292(2-3), 145-150 (2011-12-27)
The current Organisation for Economic Co-operation and Development (OECD) guidelines for evaluating organophosphorus-induced delayed neuropathy (OPIDN) require the observation of dosed animals over several days and the sacrifice of 48 hens. Adhering to these protocols in tests with enantiomers is
Teresa Rodríguez et al.
Occupational and environmental medicine, 69(2), 119-125 (2011-07-05)
This study assessed pesticide exposure of children in rural Nicaragua in relation to parental pesticide use, from around conception to current school age, as part of an epidemiological evaluation of neurodevelopment effects. We included 132 children whose parents were subsistence
Bao-Jian Hang et al.
Applied and environmental microbiology, 78(6), 1962-1968 (2012-01-17)
De-esterification is an important degradation or detoxification mechanism of sulfonylurea herbicide in microbes and plants. However, the biochemical and molecular mechanisms of sulfonylurea herbicide de-esterification are still unknown. In this study, a novel esterase gene, sulE, responsible for sulfonylurea herbicide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service