Skip to Content
Merck
All Photos(1)

Key Documents

PS349

Propyzamide

analytical standard

Synonym(s):

Pronamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11Cl2NO
CAS Number:
Molecular Weight:
256.13
Beilstein:
882391
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-349

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(NC(=O)c1cc(Cl)cc(Cl)c1)C#C

InChI

1S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7H,2-3H3,(H,15,16)

InChI key

PHNUZKMIPFFYSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Kwiatkowska et al.
Cell biology international, 36(5), 455-462 (2012-02-03)
Lipid bodies present in lipotubuloids of Ornithogalum umbellatum ovary epidermis take the form of a lens between leaflets of ER (endoplasmic reticulum) membrane filled with a highly osmiophilic substance. The two enzymes, DGAT1 [DAG (diacylglycerol) acyltransferase 1] and DGAT2 (DAG
K Yoshida et al.
Protoplasma, 221(3-4), 227-235 (2003-06-13)
Some species of Spirogyra form rosette-shaped or rod-shaped rhizoids in the terminal cell of the filaments. In the present study, we analyzed an involvement of microtubules (MTs) in rhizoid differentiation. Before rhizoid differentiation, cortical MTs were arranged transversely to the
Aaron A Santner et al.
The Plant journal : for cell and molecular biology, 45(5), 752-764 (2006-02-08)
The WAG1 and WAG2 genes of Arabidopsis thaliana encode protein-serine/threonine kinases that are closely related to PINOID. In order to determine what roles WAG1 and WAG2 play in seedling development, we used a reverse genetics approach to study the wag1
Y Makimoto et al.
Protoplasma, 229(1), 53-62 (2006-10-05)
Fibrillarin is known to play an important role in precursor ribosomal RNA processing and ribosome assembly. The present study describes a fibrillarin homolog gene isolated from tobacco BY-2 cells and its expression during the cell cycle. The cDNA for a
Hiroyasu Motose et al.
The Plant journal : for cell and molecular biology, 67(6), 993-1005 (2011-05-25)
NimA-related kinase 6 (NEK6) has been implicated in microtubule regulation to suppress the ectopic outgrowth of epidermal cells; however, its molecular functions remain to be elucidated. Here, we analyze the function of NEK6 and other members of the NEK family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service