Skip to Content
Merck
All Photos(1)

Key Documents

PS2044

Supelco

Thidiazuron

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N4OS
CAS Number:
Molecular Weight:
220.25
Beilstein:
1078092
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-2044

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(Nc1ccccc1)Nc2cnns2

InChI

1S/C9H8N4OS/c14-9(12-8-6-10-13-15-8)11-7-4-2-1-3-5-7/h1-6H,(H2,11,12,14)

InChI key

HFCYZXMHUIHAQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Cutri et al.
Mechanisms of development, 130(1), 61-69 (2012-06-05)
Tendrils can be found in different plant species. In legumes such as pea, tendrils are modified leaves produced by the vegetative meristem but in the grape vine, a same meristem is used to either form a tendril or an inflorescence.
Alexander V Babosha
Journal of plant physiology, 166(17), 1892-1903 (2009-07-14)
Dose-response relationship between resistance of wheat seedlings (Triticum aestivum, cultivar Zarya) to Erysiphe graminis f. sp. tritici Marchal. (Syn. Blumeria graminis), a causal organism of wheat powdery mildew and exogenous zeatin has been investigated. Two-week-old seedlings were inoculated with the
Hemaid Ibrahim Soliman et al.
GM crops, 1(1), 40-51 (2010-01-01)
Fig is one of the most important fruit trees in Egypt. It used to constitute the major source of income for the inhabitants of the western north coast of Egypt. Since 1993 fig cultivations were threatened by a number of
Dmytro P Yevtushenko et al.
Plant cell reports, 29(3), 211-221 (2010-01-21)
Many economically important species of Populus, especially those in sections Aigeiros and Tacamahaca, remain recalcitrant to genetic transformation. In this study, a simple and reliable protocol was developed for the efficient Agrobacterium-mediated transformation of a difficult-to-transform, but commercially viable, hybrid
M Cano-Castillo et al.
Cryo letters, 33(3), 182-190 (2012-07-25)
We cryopreserved in vitro shoot tips of saltcedar (Tamarix boveana Bunge) using the vitrification technique. The success of the cryopreservation protocol was strongly affected by preculture, loading duration, dehydration duration in plant vitrification solution 2 (PVS2), and medium composition during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service