Skip to Content
Merck
All Photos(1)

Key Documents

PS1078

Metsulfuron-methyl

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N5O6S
CAS Number:
Molecular Weight:
381.36
Beilstein:
587472
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1078

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2

InChI

1S/C14H15N5O6S/c1-8-15-12(18-14(16-8)25-3)17-13(21)19-26(22,23)10-7-5-4-6-9(10)11(20)24-2/h4-7H,1-3H3,(H2,15,16,17,18,19,21)

InChI key

RSMUVYRMZCOLBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metsulfuron-methyl is a sulfonylurea herbicide, which finds broad-spectrum of applications in weed control management in cultivation of a variety of crops.

Application

Metsulfuron-methyl may be used as a reference standard for the following:
  • Determining the metsulfuron-methyl content in soil and water samples using gas chromatography coupled with mass spectrometry (GC-MS).
  • Analysis of metsulfuron-methyl in water samples extracted using multiwalled carbon nanotubes solid phase extraction packing material by high performance liquid chromatography (HPLC).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Different toxic effects of the sulfonylurea herbicides metsulfuron methyl, chlorsulfuron and thifensulfuron methyl on fluorescent pseudomonads isolated from an agricultural soil.
Boldt S T and Jacobsen S C.
FEMS Microbiology Letters, 161(1), 29-35 (1998)
Issues in Applied, Analytical, and Imaging Sciences Research: 2013 Edition (2013)
Effects of the herbicides clomazone, quinclorac, and metsulfuron methyl on acetylcholinesterase activity in the silver catfish (Rhamdia quelen)(Heptapteridae).
Miron S D D, et al.
Ecotox. Environ. Safety, 61(3), 398-403 (2005)
Analytical procedure for the determination of Ethyl Lauroyl Arginate (LAE) to assess the kinetics and specific migration from a new antimicrobial active food packaging.
Pezo D, et al.
Analytica Chimica Acta, 745, 92-98 (2012)
Fate and behaviour of triasulfuron, metsulfuron-methyl, and chlorsulfuron in the Australian soil environment: a review.
Sarmah K A, et al.
Australian Journal of Agricultural Research, 49(5), 775-790 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service