Skip to Content
Merck
All Photos(1)

Documents

PS100

Piperonylbutoxide

analytical standard

Synonym(s):

2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
Beilstein:
288063
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-100

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.498 (lit.)

bp

155 °C/0.3 mmHg (lit.)

density

1.059 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCOCCOCCOCc1cc2OCOc2cc1CCC

InChI

1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3

InChI key

FIPWRIJSWJWJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bian Liu et al.
Environment international, 48, 156-161 (2012-09-01)
Previously we reported that airborne concentrations of cis-permethrin, but not trans-permethrin, measured during pregnancy in an inner city pediatric cohort was associated with cough by age 5. However, the effect of subsequent exposures to both permethrins during early childhood, and
Graham D Moores et al.
Pest management science, 68(5), 795-800 (2012-01-10)
Previous work has characterised pyrethroid resistance in pollen beetle (Meligethes aeneus F.) as principally an oxidative mechanism. Piperonyl butoxide (PBO) can synergise this resistance in the field, but its effects on the honey bee are thought to be unacceptable. A
Shu-Ping Wang et al.
Archives of insect biochemistry and physiology, 79(4-5), 264-275 (2012-04-18)
Methanol is one of the most common short-chain alcohols in fermenting fruits, the natural food of the fruit fly, Drosophila melanogaster. The larvae cope continuously with methanol at various concentrations in order to survive and develop. In the present article
Olivier J T Briët et al.
Malaria journal, 12, 77-77 (2013-02-28)
The effectiveness of insecticide-treated nets in preventing malaria is threatened by developing resistance against pyrethroids. Little is known about how strongly this affects the effectiveness of vector control programmes. Data from experimental hut studies on the effects of long-lasting, insecticidal
N Karatolos et al.
Insect molecular biology, 21(3), 327-334 (2012-03-31)
Spiromesifen is a novel insecticide and is classed as a tetronic acid derivative. It targets the insects' acetyl-coenzyme A carboxylase (ACCase) enzyme, causing a reduction in lipid biosynthesis. At the time of this publication, there are no reports of resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service