PHL86736
Sterubin
phyproof® Reference Substance
Synonym(s):
3′,4′,5-Trihydroxy 7-methoxyflavanone, Eriodictyol 7-methyl ether, Sternbin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
pharmaceutical
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Analysis Note
For this reference substance sterubin we use the non-stereospecific CAS number and structure. It is expected that the natural enantiomer is predominant; however, the substance is prone to racemization in solution. Therefore, the stereochemistry is not specified and we recommend to use this reference substance only for non-stereospecific analysis.
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service