Skip to Content
Merck
All Photos(1)

Key Documents

PHL82643

Resorcinol

phyproof® Reference Substance

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$2,115.00

R$2,115.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$2,115.00

About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$2,115.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

product line

phyproof® Reference Substance

Assay

≥95.0% (GC)

form

solid

autoignition temp.

1126 °F

manufacturer/tradename

PhytoLab

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: allium zingiber

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service