Skip to Content
Merck
All Photos(1)

Key Documents

PHL82559

Hexahydrocurcumin

phyproof® Reference Substance

Synonym(s):

(RS)-5-Hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)-3-heptanone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$7,634.00

R$7,634.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$7,634.00

About This Item

Empirical Formula (Hill Notation):
C21H26O6
CAS Number:
Molecular Weight:
374.43
Beilstein:
2486876
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$7,634.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O=C(CC(O)CCC1=CC(OC)=C(O)C=C1)CCC2=CC(OC)=C(O)C=C2

InChI

1S/C21H26O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,16,22,24-25H,3-4,7-8,13H2,1-2H3

InChI key

RSAHICAPUYTWHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: curcuma

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

424.4 °F

Flash Point(C)

218 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service